Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024783.3 | NP_079059.2 | 3 | 352 | ||
XM_005253138.4 | XP_005253195.1 | 2 | 797 | |||
XM_005253139.4 | XP_005253196.1 | 2 | 797 | |||
XM_005253140.4 | XP_005253197.1 | 2 | 797 | |||
XM_005253141.4 | XP_005253198.1 | 2 | 797 | |||
XM_006718327.3 | XP_006718390.1 | 2 | 797 | |||
XM_006718328.3 | XP_006718391.1 | 2 | 797 | |||
XM_011520372.2 | XP_011518674.1 | 3 | 706 | |||
XM_011520373.2 | XP_011518675.1 | 2 | 797 | |||
XM_011520374.2 | XP_011518676.1 | 2 | 797 | |||
XM_011520375.2 | XP_011518677.1 | 2 | 797 | |||
XM_011520376.2 | XP_011518678.1 | 2 | 797 | |||
XM_011520378.2 | XP_011518680.1 | 2 | 44 | |||
XM_011520379.2 | XP_011518681.1 | 2 | 44 | |||
XM_011520381.2 | XP_011518683.1 | 2 | 797 | |||
XM_017018331.1 | XP_016873820.1 | 2 | 797 | |||
XM_017018332.1 | XP_016873821.1 | 1 | 20 | |||
XM_017018333.1 | XP_016873822.1 | 2 | 797 | |||
GenBank mRNA | AK056687.1 | - | 2 | 751 | ||
AK303066.1 | - | 3 | 305 | |||
AK303875.1 | - | 3 | 345 | |||
BC036234.1 | - | 3 | 336 | 5295953 |