Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142886.1 | NP_001136358.1 | 6 | 821 | ||
NM_024794.2 | NP_079070.1 | 5 | 857 | |||
GenBank mRNA | AK026061.1 | - | 5 | 857 | ||
BC115002.1 | - | 6 | 731 | 7482856 | ||
BC132958.1 | - | 5 | 821 | 40146980 | ||
BC132960.1 | - | 5 | 821 | 40146982 |