Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BORA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286746.1 | NP_001273675.1 | 10 | 1416 | ||
NM_001286747.1 | NP_001273676.1 | 9 | 1211 | |||
NM_024808.3 | NP_079084.3 | 10 | 1318 | |||
XM_006719868.3 | XP_006719931.2 | 8 | 1175 | |||
GenBank mRNA | AK026277.1 | - | 5 | 643 | ||
AK291496.1 | - | 10 | 1144 | |||
AK298608.1 | - | 9 | 1037 | |||
BC007119.1 | - | 1 | 102 | |||
BC025367.2 | - | 10 | 1129 | 4098541 | ||
BC044213.1 | - | 9 | 942 | |||
BC056876.1 | - | 10 | 1141 | 6473892 | ||
BX648605.1 | - | 10 | 1416 | |||
EU834129.1 | - | 10 | 1224 |