Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136041.2 | NP_001129513.1 | 10 | 971 | ||
NM_024827.3 | NP_079103.2 | 10 | 1072 | |||
XM_005265491.1 | XP_005265548.1 | 10 | 1037 | |||
XM_005265492.1 | XP_005265549.1 | 7 | 879 | |||
XM_006713338.1 | XP_006713401.1 | 7 | 835 | |||
XM_011534131.1 | XP_011532433.1 | 10 | 1160 | |||
XM_011534132.2 | XP_011532434.2 | 7 | 725 | |||
XM_011534135.1 | XP_011532437.1 | 9 | 879 | |||
XM_017007234.1 | XP_016862723.1 | 7 | 923 | |||
GenBank mRNA | AK025426.1 | - | 10 | 963 | ||
AK025890.1 | - | 10 | 963 | |||
AK293223.1 | - | 10 | 904 | |||
AK299021.1 | - | 6 | 643 | |||
AL137362.1 | - | 7 | 606 | |||
AL834223.1 | - | 9 | 1287 | |||
BC009676.1 | - | 9 | 948 | 3906049 | ||
CR457341.1 | - | 6 | 573 |