Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGNBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024835.4 | NP_079111.1 | 9, 10 | 1536 | ||
XM_005257689.2 | XP_005257746.1 | 9, 10 | 1518 | |||
XM_011525265.2 | XP_011523567.1 | 8, 9 | 1281 | |||
XM_011525266.1 | XP_011523568.1 | 8, 9 | 1404 | |||
XM_017025107.1 | XP_016880596.1 | 8, 9 | 1249 | |||
GenBank mRNA | AF126964.1 | - | 8, 9 | 1267 | ||
AJ404670.2 | - | 1, 2 | 92 | |||
AK024883.1 | - | 9, 10 | 1490 | |||
AK026214.1 | - | 1, 2 | 170 | |||
AK124053.1 | - | 3, 4 | 1907 | |||
AK124084.1 | - | 3, 4 | 1452 | |||
AK290687.1 | - | 8, 9 | 1292 | |||
AK301614.1 | - | 6, 7 | 824 | |||
BC137491.1 | - | 9, 10 | 1402 | 9021491 | ||
CD557886.1 | - | 2, 3 | 366 |