Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160167.1 | NP_001153639.1 | 6 | 807 | ||
NM_001160168.1 | NP_001153640.1 | 4 | 347 | |||
NM_001160169.1 | NP_001153641.1 | 5 | 543 | |||
NM_024841.4 | NP_079117.3 | 7 | 793 | |||
GenBank mRNA | AK024275.1 | - | 4 | 294 | ||
AK055042.1 | - | 6 | 694 | |||
BX640959.1 | - | 7 | 774 | |||
DR001466.1 | - | 5 | 476 |