Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160305.2 | NP_001153777.1 | 4 | 596 | ||
NM_024860.3 | NP_079136.2 | 5 | 524 | |||
NR_134583.1 | 5 | 524 | ||||
XM_011523336.1 | XP_011521638.1 | 4 | 587 | |||
GenBank mRNA | AK024801.1 | - | 5 | 480 | ||
AK127136.1 | - | 4 | 575 | |||
AK290495.1 | - | 4 | 558 | |||
AK297288.1 | - | 4 | 699 | |||
AK304822.1 | - | 4 | 637 | |||
AK309468.1 | - | 4 | 555 | |||
BC022451.1 | - | 4 | 558 | 4794299 | ||
BC110433.1 | - | 5 | 508 | |||
BP338820.1 | - | 5 | 523 |