Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA0319L SIRNA Assays ›
Gene Symbol:
KIAA0319L
Gene Name:
KIAA0319 like
Gene Aliases:
AAVR
Chromosome Location:
Chr.1: 35433490 - 35557670 on Build GRCh38
UniGene ID:
Hs.456507
Species:
Human
siRNA ID
HSS129149
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA0319L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024874.4
NP_079150.3
11
1981
XM_006710907.1
XP_006710970.1
11
1780
XM_006710909.1
XP_006710972.1
11
1872
XM_006710910.1
XP_006710973.1
10
1424
XM_011542179.2
XP_011540481.1
11
2112
XM_011542180.1
XP_011540482.1
11
1950
XM_017002366.1
XP_016857855.1
11
1950
XM_017002367.1
XP_016857856.1
11
2112
XM_017002368.1
XP_016857857.1
11
1780
XM_017002369.1
XP_016857858.1
11
1872
XM_017002370.1
XP_016857859.1
11
1950
XM_017002371.1
XP_016857860.1
11
1950
XM_017002372.1
XP_016857861.1
11
1872
XM_017002373.1
XP_016857862.1
11
1950
XM_017002374.1
XP_016857863.1
10
1424
XM_017002375.1
XP_016857864.1
8
1066
XM_017002376.1
XP_016857865.1
8
1066
XM_017002377.1
XP_016857866.1
10
1424
XM_017002378.1
XP_016857867.1
2
90
GenBank mRNA
AB058740.1
-
6
679
AF275679.2
-
11
1930
AF289597.1
-
9
1546
AK024287.1
-
7
744
AK090878.1
-
2
91
AK314605.1
-
11
1929
AL834315.1
-
7
815
AY163234.1
-
11
1930
BC014530.1
-
11
1913
3843301
BC031672.1
-
6
675
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline