Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142555.2 | NP_001136027.1 | 6, 7 | 507 | ||
NM_024876.3 | NP_079152.3 | 7, 8 | 875 | |||
GenBank mRNA | BC013114.1 | - | 7, 8 | 685 | 4342891 | |
BC027473.1 | - | 6, 7 | 474 | 4997823 |