Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNDIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323606.1 | NP_001310535.1 | 3 | 1915 | ||
NM_001323607.1 | NP_001310536.1 | 2 | 591 | |||
NM_024893.2 | NP_079169.1 | 2 | 1026 | |||
XM_011529347.2 | XP_011527649.1 | 2 | 844 | |||
XM_011529348.2 | XP_011527650.1 | 2 | 844 | |||
XM_011529349.2 | XP_011527651.1 | 2 | 844 | |||
XM_011529350.1 | XP_011527652.1 | 3 | 1480 | |||
XM_011529351.1 | XP_011527653.1 | 3 | 1387 | |||
XM_011529352.2 | XP_011527654.1 | 2 | 585 | |||
XM_011529353.2 | XP_011527655.1 | 2 | 602 | |||
XM_011529354.2 | XP_011527656.1 | 2 | 582 | |||
XM_011529356.2 | XP_011527658.1 | 2 | 844 | |||
XM_011529358.2 | XP_011527660.1 | 2 | 844 | |||
XM_017028064.1 | XP_016883553.1 | 2 | 3861 | |||
XM_017028065.1 | XP_016883554.1 | 3 | 1387 | |||
XM_017028066.1 | XP_016883555.1 | 2 | 744 | |||
XM_017028067.1 | XP_016883556.1 | 2 | 650 | |||
XM_017028068.1 | XP_016883557.1 | 2 | 1611 | |||
XM_017028069.1 | XP_016883558.1 | 2 | 844 | |||
GenBank mRNA | AK024282.1 | - | 2 | 1026 | ||
BC030637.1 | - | 2 | 528 | 4821088 | ||
CR457325.1 | - | 1 | 393 |