Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024896.2 | NP_079172.2 | 13 | 2405 | ||
XM_005251587.1 | XP_005251644.1 | 11 | 2010 | |||
XM_017015139.1 | XP_016870628.1 | 14 | 2198 | |||
GenBank mRNA | AB058718.2 | - | 13 | 2282 | ||
AK026962.1 | - | 5 | 711 | |||
AK093217.1 | - | 11 | 1581 | |||
AK127218.1 | - | 13 | 2394 | |||
AK172831.1 | - | 7 | 1109 | |||
AL834191.1 | - | 12 | 2037 | |||
BC031630.1 | - | 5 | 769 | |||
BC136771.1 | - | 13 | 2379 | 9020761 | ||
BC136773.1 | - | 13 | 2379 | 9020763 | ||
BC144404.1 | - | 13 | 2379 |