Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290331.1 | NP_001277260.1 | 20 | 1675 | ||
NM_024898.3 | NP_079174.2 | 22 | 1784 | |||
XM_006722905.2 | XP_006722968.2 | 20 | 1528 | |||
XM_006722906.3 | XP_006722969.1 | 22 | 1713 | |||
XM_011528318.2 | XP_011526620.1 | 21 | 1704 | |||
GenBank mRNA | AK026410.1 | - | 22 | 1763 | ||
AK055820.1 | - | 11 | 1391 | |||
AK225623.1 | - | 22 | 1763 | |||
AK303226.1 | - | 20 | 1654 | |||
AL713770.1 | - | 20 | 1904 | |||
AL834293.1 | - | 5 | 385 | |||
BC033437.1 | - | 22 | 1751 | 5187180 | ||
BX647304.1 | - | 10 | 1414 |