Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271989.1 | NP_001258918.1 | 9 | 1353 | ||
NM_024899.3 | NP_079175.2 | 10 | 1578 | |||
NR_073537.1 | 10 | 1573 | ||||
XM_005258149.4 | XP_005258206.1 | 10 | 1557 | |||
XM_017025981.1 | XP_016881470.1 | 10 | 1557 | |||
XM_017025982.1 | XP_016881471.1 | 10 | 1638 | |||
XM_017025983.1 | XP_016881472.1 | 9 | 1428 | |||
XM_017025984.1 | XP_016881473.1 | 10 | 1540 | |||
XM_017025985.1 | XP_016881474.1 | 10 | 1506 | |||
XM_017025986.1 | XP_016881475.1 | 9 | 1315 | |||
XM_017025987.1 | XP_016881476.1 | 9 | 1282 | |||
GenBank mRNA | AK022604.1 | - | 10 | 1440 | ||
AK303117.1 | - | 9 | 1300 | |||
BC026307.1 | - | 10 | 1505 | 4792211 | ||
BC033026.1 | - | 10 | 1440 |