Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037582.2 | NP_001032671.2 | 1 | 470 | ||
NM_024906.2 | NP_079182.2 | 1 | 470 | |||
GenBank mRNA | AF389338.1 | - | 1 | 385 | ||
AK024685.1 | - | 1 | 385 | |||
BC004936.1 | - | 1 | 369 | |||
BC137429.1 | - | 1 | 374 | 9021427 | ||
BC137432.1 | - | 1 | 374 | 9021430 |