Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167941.1 | NP_001161413.1 | 9 | 1057 | ||
NM_024908.3 | NP_079184.2 | 9 | 1167 | |||
XM_005254680.3 | XP_005254737.1 | 9 | 1130 | |||
GenBank mRNA | AK023035.1 | - | 9 | 1072 | ||
AK290933.1 | - | 9 | 1089 | |||
BC025247.1 | - | 9 | 1098 | 4560352 | ||
BC051855.1 | - | 9 | 1070 | 6141090 | ||
EF011624.1 | - | 9 | 1070 |