Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002292.3 | NP_001002292.3 | 1 | 309 | ||
NM_001193334.1 | NP_001180263.1 | 1 | 309 | |||
NM_024911.6 | NP_079187.3 | 1 | 364 | |||
XM_011542191.2 | XP_011540493.1 | 1 | 287 | |||
GenBank mRNA | AB097018.1 | - | 1 | 286 | ||
AK074583.1 | - | 1 | 319 | |||
AK074984.1 | - | 1 | 286 | |||
AK301613.1 | - | 1 | 286 | |||
AK309779.1 | - | 1 | 320 | |||
AY359035.1 | - | 1 | 346 | |||
BC110826.1 | - | 1 | 265 | 6183768 | ||
BC137109.1 | - | 1 | 161 | 9021106 | ||
BC137113.1 | - | 1 | 161 | 9021110 | ||
BG701224.1 | - | 1 | 338 | |||
BX538320.1 | - | 1 | 309 | |||
BX648748.1 | - | 1 | 307 | |||
DB478499.1 | - | 2 | 391 | |||
DQ323735.1 | - | 1 | 333 |