Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105533.1 | NP_001099003.1 | 6, 7 | 1146 | ||
NM_024913.4 | NP_079189.4 | 7, 8 | 1362 | |||
XM_017012649.1 | XP_016868138.1 | 6, 7 | 1838 | |||
GenBank mRNA | AK025639.1 | - | 4, 5 | 507 | ||
AK055624.1 | - | 6, 7 | 1486 | |||
AK225313.1 | - | 4, 5 | 507 | |||
AK289978.1 | - | 7, 8 | 1359 | |||
AL832619.2 | - | 7, 8 | 1317 | |||
AL832638.2 | - | 7, 8 | 1186 | |||
AY358220.1 | - | 6, 7 | 938 | |||
BC017764.1 | - | 3, 4 | 359 | |||
BC018685.1 | - | 6, 7 | 897 | |||
BC030538.2 | - | 3, 4 | 376 | 3919732 |