Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024927.4 | NP_079203.3 | 6 | 1155 | ||
NR_073573.1 | 6 | 1155 | ||||
NR_073574.1 | 6 | 716 | ||||
XM_017025113.1 | XP_016880602.1 | 7 | 1126 | |||
XM_017025114.1 | XP_016880603.1 | 7 | 1126 | |||
XM_017025115.1 | XP_016880604.1 | 7 | 1276 | |||
XM_017025116.1 | XP_016880605.1 | 6 | 991 | |||
XM_017025117.1 | XP_016880606.1 | 7 | 1126 | |||
XM_017025118.1 | XP_016880607.1 | 6 | 1141 | |||
GenBank mRNA | AB208939.1 | - | 6 | 1022 | ||
AK024672.1 | - | 6 | 783 | |||
AK096767.1 | - | 6 | 716 | |||
BC016062.1 | - | 6 | 758 | 4909623 | ||
BC052978.1 | - | 6 | 1076 | 6473233 |