Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GREB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142966.1 | NP_001136438.1 | 12 | 1930 | ||
XM_006722547.2 | XP_006722610.1 | 12 | 1893 | |||
XM_011526179.2 | XP_011524481.1 | 12 | 1893 | |||
XM_017025988.1 | XP_016881477.1 | 12 | 1893 | |||
XM_017025989.1 | XP_016881478.1 | 12 | 1966 | |||
XM_017025990.1 | XP_016881479.1 | 12 | 1893 | |||
XM_017025991.1 | XP_016881480.1 | 12 | 1893 | |||
XM_017025992.1 | XP_016881481.1 | 11 | 1764 | |||
XM_017025993.1 | XP_016881482.1 | 11 | 1837 | |||
GenBank mRNA | AB051559.1 | - | 12 | 1930 | ||
AK309723.1 | - | 11 | 1764 |