Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024940.7 | NP_079216.4 | 13 | 1479 | ||
XM_011544661.1 | XP_011542963.1 | 13 | 1416 | |||
XM_011544662.1 | XP_011542964.1 | 13 | 1416 | |||
XM_011544663.2 | XP_011542965.1 | 13 | 1416 | |||
XM_017013865.1 | XP_016869354.1 | 13 | 1268 | |||
XM_017013866.1 | XP_016869355.1 | 11 | 2373 | |||
XM_017013867.1 | XP_016869356.1 | 6 | 806 | |||
XM_017013868.1 | XP_016869357.1 | 7 | 754 | |||
GenBank mRNA | AK123607.1 | - | 13 | 1416 | ||
BC137175.1 | - | 13 | 1337 | 9021172 | ||
BC137176.1 | - | 13 | 1337 | 9021173 | ||
BM450222.1 | - | 2 | 214 | |||
CR749353.1 | - | 5 | 620 | |||
CR933718.1 | - | 1 | 53 |