Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308116.1 | NP_001295045.1 | 13, 14 | 2651 | ||
NM_024947.3 | NP_079223.3 | 13, 14 | 2636 | |||
XM_005247789.2 | XP_005247846.1 | 14, 15 | 2714 | |||
XM_006713753.1 | XP_006713816.1 | 14, 15 | 2683 | |||
XM_006713754.3 | XP_006713817.1 | 14, 15 | 2805 | |||
XM_006713756.2 | XP_006713819.1 | 12, 13 | 2456 | |||
XM_011513186.2 | XP_011511488.1 | 13, 14 | 2615 | |||
XM_011513187.2 | XP_011511489.1 | 12, 13 | 2567 | |||
XM_011513188.2 | XP_011511490.1 | 13, 14 | 2555 | |||
XM_017007235.1 | XP_016862724.1 | 14, 15 | 2702 | |||
XM_017007236.1 | XP_016862725.1 | 14, 15 | 2690 | |||
XM_017007237.1 | XP_016862726.1 | 12, 13 | 2507 | |||
XM_017007238.1 | XP_016862727.1 | 11, 12 | 2408 | |||
XM_017007239.1 | XP_016862728.1 | 11, 12 | 2255 | |||
GenBank mRNA | AF380154.1 | - | 13, 14 | 2573 | ||
AF444193.1 | - | 12, 13 | 2588 | |||
AJ320486.3 | - | 13, 14 | 2630 | |||
BC131772.1 | - | 13, 14 | 2615 | 40123356 | ||
EF560717.1 | - | 13, 14 | 2611 |