Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024953.3 | NP_079229.2 | 5 | 520 | ||
XM_005253938.1 | XP_005253995.1 | 4 | 366 | |||
XM_006719606.2 | XP_006719669.1 | 5 | 1180 | |||
XM_017019977.1 | XP_016875466.1 | 3 | 243 | |||
GenBank mRNA | AB054990.1 | - | 5 | 462 | ||
AK023151.1 | - | 2 | 165 | |||
AK292485.1 | - | 5 | 493 | |||
AK307494.1 | - | 6 | 669 | |||
AK309772.1 | - | 4 | 346 | |||
AV760516.2 | - | 5 | 485 | |||
BC034357.1 | - | 5 | 474 | |||
BC065481.1 | - | 5 | 453 | |||
BC113585.1 | - | 5 | 520 | 8322637 | ||
BC113587.1 | - | 5 | 520 | 8322639 | ||
BX641140.1 | - | 5 | 475 | |||
BX647450.1 | - | 5 | 473 | |||
BX647992.1 | - | 2 | 121 |