Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321213.1 | NP_001308142.1 | 3 | 1080 | ||
NM_024963.5 | NP_079239.3 | 3 | 1080 | |||
XM_011515531.2 | XP_011513833.1 | 3 | 1095 | |||
XM_011515532.2 | XP_011513834.1 | 3 | 1095 | |||
GenBank mRNA | AK057042.1 | - | 3 | 1034 | ||
AK095394.1 | - | 1 | 607 | |||
AK294949.1 | - | 3 | 1074 | |||
BC006426.1 | - | 3 | 1050 | 4026501 | ||
BC024256.1 | - | 3 | 1050 | 3954975 |