Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172173.1 | NP_001165644.1 | 7 | 2039 | ||
NM_024969.3 | NP_079245.2 | 5 | 1938 | |||
XM_005246865.4 | XP_005246922.1 | 5 | 1761 | |||
GenBank mRNA | AB063300.1 | - | 5 | 1999 | ||
AK021765.1 | - | 2 | 1072 | |||
AK056655.1 | - | 5 | 1935 | |||
AK126129.1 | - | 8 | 1828 |