Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
TRPM3
Gene Name:
transient receptor potential cation channel subfamily M member 3
Gene Aliases:
GON-2, LTRPC3, MLSN2
Chromosome Location:
Chr.9: 70529063 - 71446989 on Build GRCh38
UniGene ID:
Hs.47288
Species:
Human
siRNA ID
HSS129304
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007470.1
NP_001007471.1
5
632
NM_001007471.2
NP_001007472.2
5
980
NM_020952.4
NP_066003.3
5
632
NM_024971.5
NP_079247.5
5
632
NM_206944.3
NP_996827.3
5
632
NM_206945.3
NP_996828.3
5
632
NM_206946.3
NP_996829.3
5
632
NM_206947.3
NP_996830.3
5
632
NM_206948.2
NP_996831.1
5
632
XM_011519035.2
XP_011517337.1
5
857
XM_011519036.2
XP_011517338.1
5
3782
XM_011519037.2
XP_011517339.1
5
3768
XM_011519038.2
XP_011517340.1
5
3765
XM_011519039.2
XP_011517341.1
5
3766
XM_011519040.2
XP_011517342.1
5
863
XM_011519041.2
XP_011517343.1
5
3765
XM_011519042.2
XP_011517344.1
5
863
XM_011519043.2
XP_011517345.1
5
896
XM_011519044.2
XP_011517346.1
5
3766
XM_011519045.2
XP_011517347.1
5
857
XM_011519046.2
XP_011517348.1
5
2560
XM_011519047.2
XP_011517349.1
5
2530
XM_017015141.1
XP_016870630.1
5
857
XM_017015142.1
XP_016870631.1
5
857
XM_017015143.1
XP_016870632.1
5
857
XM_017015144.1
XP_016870633.1
5
623
XM_017015145.1
XP_016870634.1
5
1301
XM_017015146.1
XP_016870635.1
5
699
XM_017015147.1
XP_016870636.1
5
2734
XM_017015148.1
XP_016870637.1
5
857
XM_017015149.1
XP_016870638.1
5
857
XM_017015150.1
XP_016870639.1
5
857
XM_017015151.1
XP_016870640.1
5
875
XM_017015152.1
XP_016870641.1
5
2535
XM_017015153.1
XP_016870642.1
5
867
XM_017015154.1
XP_016870643.1
5
1003
XM_017015155.1
XP_016870644.1
5
2532
XM_017015156.1
XP_016870645.1
1
55
XM_017015157.1
XP_016870646.1
5
2530
XM_017015158.1
XP_016870647.1
5
2528
XM_017015159.1
XP_016870648.1
5
623
XM_017015160.1
XP_016870649.1
5
713
XM_017015161.1
XP_016870650.1
5
857
GenBank mRNA
AB099661.1
-
2
277
AB099662.1
-
2
277
AB099663.1
-
2
277
AB099664.1
-
2
277
AB099665.1
-
2
277
AF325212.1
-
5
603
AF536748.1
-
2
277
AF536749.1
-
2
277
AF536750.1
-
2
277
AF536751.1
-
2
277
AF536752.1
-
2
277
AF536753.1
-
2
277
AJ505025.1
-
5
980
AJ505026.1
-
5
980
AK021788.1
-
5
623
AK225732.1
-
5
623
AK308682.1
-
5
808
AL136545.1
-
5
813
BC022454.2
-
4
562
BC067733.1
-
5
594
BC094699.1
-
5
626
BC121821.1
-
3
478
BC134414.1
-
3
481
BC142972.1
-
5
649
KF987075.1
-
5
688
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
transporter
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
detection of temperature stimulus
sensory perception of temperature stimulus
protein tetramerization
calcium ion transmembrane transport
calcium activated cation channel activity
cation channel activity
calcium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0