Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025059.3 | NP_079335.2 | 4 | 655 | ||
XM_011536147.2 | XP_011534449.1 | 4 | 693 | |||
XM_011536148.2 | XP_011534450.1 | 4 | 696 | |||
GenBank mRNA | AK026958.1 | - | 4 | 568 | ||
AK225330.1 | - | 4 | 569 | |||
BC060803.1 | - | 4 | 604 | 5298914 | ||
BI562667.1 | - | 4 | 609 |