Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIKE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102396.1 | NP_001095866.1 | 3 | 413 | ||
NM_025073.2 | NP_079349.2 | 3 | 401 | |||
NR_049741.1 | 3 | 463 | ||||
NR_049742.1 | 2 | 295 | ||||
GenBank mRNA | AI221103.1 | - | 2 | 120 | ||
AK024821.1 | - | 3 | 391 | |||
AK126316.1 | - | 3 | 411 | |||
AK225734.1 | - | 3 | 392 | |||
BC005934.1 | - | 3 | 371 | 4249034 | ||
CR749219.1 | - | 3 | 404 | |||
CR936771.1 | - | 3 | 404 | |||
DB459255.1 | - | 3 | 231 |