Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UXS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253875.1 | NP_001240804.1 | 15 | 1289 | ||
NM_001253876.1 | NP_001240805.1 | 10 | 1426 | |||
NM_025076.4 | NP_079352.2 | 15 | 1274 | |||
NR_045607.1 | 14 | 1225 | ||||
XM_006712775.3 | XP_006712838.1 | 14 | 1147 | |||
XM_011511902.2 | XP_011510204.2 | 15 | 1268 | |||
XM_011511903.2 | XP_011510205.2 | 15 | 1251 | |||
XM_011511904.2 | XP_011510206.1 | 12 | 1596 | |||
XM_017005008.1 | XP_016860497.1 | 13 | 1089 | |||
XM_017005009.1 | XP_016860498.1 | 13 | 1074 | |||
XM_017005010.1 | XP_016860499.1 | 14 | 1132 | |||
XM_017005011.1 | XP_016860500.1 | 15 | 1376 | |||
XM_017005012.1 | XP_016860501.1 | 15 | 1311 | |||
XM_017005013.1 | XP_016860502.1 | 13 | 1834 | |||
XM_017005014.1 | XP_016860503.1 | 14 | 1052 | |||
GenBank mRNA | AK027244.1 | - | 10 | 1426 | ||
AK075120.1 | - | 15 | 1236 | |||
AK075170.1 | - | 15 | 1251 | |||
AK122696.1 | - | 14 | 1190 | |||
AK225592.1 | - | 10 | 1426 | |||
AK290668.1 | - | 15 | 1249 | |||
AL834439.1 | - | 2 | 1038 | |||
AY147934.1 | - | 15 | 1241 | |||
AY358541.1 | - | 15 | 1225 | |||
BC009819.2 | - | 15 | 1215 | 3843312 |