Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025107.2 | NP_079383.2 | 2 | 369 | ||
XM_011536149.2 | XP_011534451.1 | 2 | 369 | |||
XM_011536150.2 | XP_011534452.1 | 2 | 369 | |||
XM_011536151.2 | XP_011534453.1 | 2 | 369 | |||
XM_011536152.2 | XP_011534454.1 | 2 | 369 | |||
XM_017011319.1 | XP_016866808.1 | 2 | 369 | |||
GenBank mRNA | AF527367.1 | - | 2 | 363 | ||
AK024922.1 | - | 2 | 361 | |||
AL834500.1 | - | 2 | 268 | |||
BC022556.1 | - | 2 | 318 | |||
BC128463.1 | - | 2 | 361 | 40126671 | ||
BC128464.1 | - | 2 | 361 | 40126673 | ||
DA863771.1 | - | 2 | 369 | |||
GU997693.1 | - | 2 | 357 |