Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102401.2 | NP_001095871.1 | 6 | 1333 | ||
NM_001265581.1 | NP_001252510.1 | 6 | 1455 | |||
NM_025115.3 | NP_079391.2 | 5 | 1702 | |||
XM_011544665.2 | XP_011542967.1 | 5 | 1656 | |||
GenBank mRNA | AK026916.1 | - | 5 | 1555 | ||
BC007387.2 | - | 6 | 1258 | 3830500 | ||
BC066935.1 | - | 6 | 1286 | 4839169 |