Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160227.1 | NP_001153699.1 | 10, 11 | 2106 | ||
NM_025137.3 | NP_079413.3 | 10, 11 | 2106 | |||
XM_006720700.1 | XP_006720763.1 | 10, 11 | 2106 | |||
XM_006720701.3 | XP_006720764.1 | 10, 11 | 2106 | |||
XM_017022634.1 | XP_016878123.1 | 10, 11 | 2106 | |||
XM_017022635.1 | XP_016878124.1 | 10, 11 | 2106 | |||
XM_017022636.1 | XP_016878125.1 | 22 | ||||
GenBank mRNA | AB058743.2 | - | 10, 11 | 2076 | ||
AB470308.1 | - | 10, 11 | 2075 | |||
AB470309.1 | - | 10, 11 | 2075 | |||
AB470310.1 | - | 10, 11 | 2075 | |||
AL834168.1 | - | 9, 10 | 1828 | |||
BC047416.1 | - | 10, 11 | 2086 | |||
BC150640.1 | - | 10, 11 | 2103 | 9057010 | ||
BC153879.1 | - | 10, 11 | 2076 | 8860108 |