Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160227.1 | NP_001153699.1 | 38 | 6990 | ||
NM_025137.3 | NP_079413.3 | 40 | 7329 | |||
XM_006720700.1 | XP_006720763.1 | 40 | 7185 | |||
XM_017022634.1 | XP_016878123.1 | 39 | 7221 | |||
XM_017022636.1 | XP_016878125.1 | 29 | 5138 | |||
GenBank mRNA | AB058743.2 | - | 40 | 7299 | ||
AB470308.1 | - | 37 | 6901 | |||
AB470309.1 | - | 40 | 7298 | |||
AB470310.1 | - | 40 | 7298 | |||
AK025092.1 | - | 11 | 1450 | |||
AK057869.1 | - | 8 | 1669 | |||
AK225186.1 | - | 11 | 1450 | |||
AY954502.1 | - | 1 | 168 | |||
BC024161.1 | - | 10 | 1415 | |||
BC067798.1 | - | 11 | 1691 | |||
BC094704.1 | - | 11 | 1823 | |||
BC150640.1 | - | 38 | 6987 | 9057010 | ||
BC153879.1 | - | 40 | 7299 | 8860108 |