Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025138.4 | NP_079414.3 | 11 | 3011 | ||
XM_005266545.3 | XP_005266602.1 | 10 | 2984 | |||
XM_011535239.2 | XP_011533541.1 | 10 | 2944 | |||
GenBank mRNA | AB107354.1 | - | 11 | 3011 | ||
AK022723.1 | - | 5 | 1687 | |||
AK074360.1 | - | 11 | 2983 | |||
AK299663.1 | - | 8 | 2007 | |||
AL833854.1 | - | 1 | 738 | |||
AL834435.1 | - | 1 | 227 | |||
BC053566.1 | - | 5 | 1690 | |||
BC064348.1 | - | 11 | 2266 | 4939198 | ||
BX647471.1 | - | 11 | 2433 |