Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025145.5 | NP_079421.5 | 29 | 3828 | ||
XM_005270171.2 | XP_005270228.1 | 29 | 3832 | |||
XM_005270172.3 | XP_005270229.1 | 29 | 3832 | |||
XM_011540196.2 | XP_011538498.1 | 30 | 3946 | |||
XM_011540197.2 | XP_011538499.1 | 30 | 3946 | |||
XM_011540198.2 | XP_011538500.1 | 29 | 3829 | |||
XM_011540199.2 | XP_011538501.1 | 29 | 3829 | |||
XM_011540201.2 | XP_011538503.1 | 30 | 3946 | |||
XM_011540202.2 | XP_011538504.1 | 26 | 3262 | |||
XM_011540203.2 | XP_011538505.1 | 28 | 3504 | |||
XM_017016681.1 | XP_016872170.1 | 30 | 3943 | |||
XM_017016682.1 | XP_016872171.1 | 27 | 3598 | |||
XM_017016683.1 | XP_016872172.1 | 27 | 3384 | |||
GenBank mRNA | AK093325.1 | - | 3 | 258 | ||
AL833836.1 | - | 15 | 1794 | |||
DA681478.1 | - | 4 | 384 |