Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288968.1 | NP_001275897.1 | 6 | 722 | ||
NM_001288969.1 | NP_001275898.1 | 5 | 608 | |||
NM_001288970.1 | NP_001275899.1 | 4 | 518 | |||
NM_001288971.1 | NP_001275900.1 | 3 | 322 | |||
NM_001288972.1 | NP_001275901.1 | 5 | 564 | |||
NM_025149.5 | NP_079425.3 | 5 | 647 | |||
XM_006722110.3 | XP_006722173.1 | 5 | 587 | |||
XM_011525294.1 | XP_011523596.1 | 5 | 587 | |||
GenBank mRNA | AK024573.1 | - | 5 | 554 | ||
AK225044.1 | - | 5 | 554 | |||
AK225561.1 | - | 5 | 553 | |||
AK294205.1 | - | 6 | 664 | |||
AK295258.1 | - | 5 | 516 | |||
AK300145.1 | - | 4 | 426 | |||
AK300625.1 | - | 3 | 322 | |||
AY358660.1 | - | 5 | 583 | |||
BC012053.1 | - | 5 | 560 | 4577089 | ||
BC014123.2 | - | 5 | 567 | 4560029 | ||
DC296100.1 | - | 5 | 504 | |||
DC384989.1 | - | 3 | 322 |