Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271895.1 | NP_001258824.1 | 8 | 955 | ||
NM_001271896.1 | NP_001258825.1 | 6 | 811 | |||
NM_025150.4 | NP_079426.2 | 10 | 1201 | |||
NR_073513.1 | 6 | 600 | ||||
NR_073514.1 | 9 | 1076 | ||||
XM_006711555.1 | XP_006711618.1 | 10 | 1164 | |||
XM_006711556.1 | XP_006711619.1 | 9 | 1017 | |||
XM_017002394.1 | XP_016857883.1 | 9 | 1017 | |||
XM_017002395.1 | XP_016857884.1 | 8 | 918 | |||
GenBank mRNA | AK022590.1 | - | 6 | 761 | ||
AK222814.1 | - | 10 | 1142 | |||
AK225368.1 | - | 8 | 875 | |||
AK298773.1 | - | 9 | 1029 | |||
AK315507.1 | - | 8 | 901 | |||
AY450558.1 | - | 6 | 530 | |||
BC000541.2 | - | 10 | 1148 | 3163506 | ||
BC007824.2 | - | 10 | 1089 | |||
BC009997.2 | - | 10 | 1121 | 4128881 | ||
BP308698.1 | - | 2 | 118 |