Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201573.1 | NP_001188502.1 | 5 | 351 | ||
NM_001201574.1 | NP_001188503.1 | 2 | 254 | |||
NM_025152.2 | NP_079428.2 | 7 | 587 | |||
XM_011537181.2 | XP_011535483.1 | 4 | 522 | |||
XM_011537182.2 | XP_011535484.1 | 8 | 561 | |||
XM_011537184.2 | XP_011535486.1 | 2 | 437 | |||
XM_017021667.1 | XP_016877156.1 | 6 | 508 | |||
GenBank mRNA | AK022722.1 | - | 7 | 500 | ||
AK093789.1 | - | 2 | 254 | |||
AK295326.1 | - | 7 | 548 | |||
AK301447.1 | - | 5 | 351 | |||
AK316445.1 | - | 7 | 577 | |||
BC024919.1 | - | 7 | 513 | 4475196 | ||
KJ534907.1 | - | 7 | 532 | |||
KJ534908.1 | - | 7 | 532 | |||
KJ534909.1 | - | 7 | 532 |