Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115113.2 | NP_001108585.2 | 4 | 798 | ||
NM_025160.6 | NP_079436.4 | 4 | 846 | |||
XM_005273273.4 | XP_005273330.1 | 4 | 747 | |||
XM_006711817.3 | XP_006711880.1 | 4 | 747 | |||
XM_011544281.2 | XP_011542583.1 | 4 | 587 | |||
XM_017002396.1 | XP_016857885.1 | 4 | 435 | |||
GenBank mRNA | AB209887.1 | - | 4 | 846 | ||
AK299230.1 | - | 1 | 31 | |||
AK307909.1 | - | 4 | 512 | |||
AY221751.1 | - | 4 | 636 | |||
AY304473.1 | - | 4 | 403 | |||
BC034498.1 | - | 4 | 649 | |||
BC052301.1 | - | 3 | 242 | 6420398 | ||
BG744504.1 | - | 4 | 510 | |||
CT002105.1 | - | 4 | 577 | |||
EF011612.1 | - | 3 | 241 |