Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM214B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317991.1 | NP_001304920.1 | 8 | 1667 | ||
NM_025182.3 | NP_079458.2 | 8 | 1668 | |||
NR_134455.1 | 9 | 1304 | ||||
XM_005251588.1 | XP_005251645.1 | 8 | 1802 | |||
XM_005251590.1 | XP_005251647.1 | 8 | 1605 | |||
XM_005251591.1 | XP_005251648.1 | 8 | 1641 | |||
XM_005251592.1 | XP_005251649.1 | 8 | 1082 | |||
XM_005251593.1 | XP_005251650.1 | 8 | 964 | |||
XM_005251594.1 | XP_005251651.1 | 8 | 838 | |||
XM_005251596.4 | XP_005251653.1 | 8 | 826 | |||
XM_005251597.4 | XP_005251654.1 | 8 | 869 | |||
XM_005251598.4 | XP_005251655.1 | 8 | 804 | |||
XM_011518037.1 | XP_011516339.1 | 8 | 1624 | |||
XM_011518039.1 | XP_011516341.1 | 7 | 1733 | |||
XM_011518040.1 | XP_011516342.1 | 9 | 1688 | |||
XM_011518043.1 | XP_011516345.1 | 9 | 1106 | |||
XM_011518044.1 | XP_011516346.1 | 9 | 1026 | |||
XM_017015169.1 | XP_016870658.1 | 9 | 1631 | |||
XM_017015170.1 | XP_016870659.1 | 9 | 1322 | |||
GenBank mRNA | AB040972.1 | - | 7 | 4398 | ||
AK022837.1 | - | 9 | 1182 | |||
AL583909.1 | - | 8 | 1667 | |||
BC004406.1 | - | 8 | 1653 | 3638496 |