Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243252.1 | NP_001230181.1 | 3 | 370 | ||
NM_001308395.1 | NP_001295324.1 | 4 | 842 | |||
NM_025202.3 | NP_079478.1 | 3 | 763 | |||
XM_011511924.2 | XP_011510226.1 | 3 | 611 | |||
GenBank mRNA | AF193044.1 | - | 2 | 307 | ||
AF218006.1 | - | 3 | 638 | |||
AK293581.1 | - | 4 | 722 | |||
AK315442.1 | - | 3 | 703 | |||
BC002449.2 | - | 3 | 646 | 3343900 | ||
BI460146.1 | - | 4 | 633 | |||
CR627385.1 | - | 3 | 596 | |||
DB461847.1 | - | 3 | 367 |