Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243252.1 | NP_001230181.1 | 4 | 412 | ||
NM_001308395.1 | NP_001295324.1 | 5 | 884 | |||
NM_025202.3 | NP_079478.1 | 4 | 805 | |||
XM_011511924.2 | XP_011510226.1 | 4 | 653 | |||
GenBank mRNA | AB209715.1 | - | 4 | 676 | ||
AF176923.1 | - | 4 | 345 | |||
AF193044.1 | - | 3 | 349 | |||
AF218006.1 | - | 4 | 680 | |||
AK023674.1 | - | 4 | 697 | |||
AK293581.1 | - | 5 | 764 | |||
AK310565.1 | - | 4 | 412 | |||
AK315442.1 | - | 4 | 745 | |||
BC002449.2 | - | 4 | 688 | 3343900 | ||
BC004128.2 | - | 4 | 676 | 3940060 | ||
BC035476.1 | - | 4 | 744 | |||
BI460146.1 | - | 5 | 675 | |||
CR627385.1 | - | 4 | 638 | |||
DB461847.1 | - | 4 | 409 |