Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025207.4 | NP_079483.3 | 6, 7 | 2025 | ||
NM_201398.2 | NP_958800.1 | 7, 8 | 1581 | |||
GenBank mRNA | AF481877.1 | - | 6, 7 | 1333 | ||
AF520568.1 | - | 6, 7 | 1946 | |||
AK025979.1 | - | 7, 8 | 1509 | |||
BC020253.1 | - | 5, 6 | 860 | |||
BC021096.1 | - | 7, 8 | 1499 | 4891331 | ||
BC032323.1 | - | 7, 8 | 1516 | 5242727 | ||
DQ458779.1 | - | 6, 7 | 1624 | |||
U79241.1 | - | 5, 6 | 1176 |