Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308189.1 | NP_001295118.1 | 1 | 2396 | ||
NM_001308191.1 | NP_001295120.1 | 1 | 2396 | |||
NM_030627.3 | NP_085130.2 | 1 | 2396 | |||
XM_005265994.1 | XP_005266051.1 | 1 | 2396 | |||
XM_011534660.2 | XP_011532962.1 | 1 | 2396 | |||
XM_011534661.2 | XP_011532963.1 | 1 | 2396 | |||
GenBank mRNA | AB051460.1 | - | 1 | 1058 | ||
AK310905.1 | - | 1 | 1193 | |||
BC117150.1 | - | 1 | 1384 | 40125701 | ||
BC143958.1 | - | 1 | 1384 | 9052476 | ||
BC143959.1 | - | 1 | 1384 | 9052477 | ||
BX538213.1 | - | 1 | 1538 |