Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308189.1 | NP_001295118.1 | 8, 9 | 3326 | ||
NM_001308191.1 | NP_001295120.1 | 7, 8 | 3302 | |||
NM_001308192.1 | NP_001295121.1 | 8, 9 | 955 | |||
NM_001308193.1 | NP_001295122.1 | 7, 8 | 904 | |||
NM_030627.3 | NP_085130.2 | 9, 10 | 3377 | |||
XM_005265994.1 | XP_005266051.1 | 8, 9 | 3353 | |||
GenBank mRNA | AB051460.1 | - | 8, 9 | 1988 | ||
BC036899.1 | - | 7, 8 | 888 | |||
BC117150.1 | - | 9, 10 | 2365 | 40125701 | ||
BC143958.1 | - | 8, 9 | 2314 | 9052476 | ||
BC143959.1 | - | 7, 8 | 2290 | 9052477 | ||
BX538213.1 | - | 9, 10 | 2519 |