Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP70 SIRNA Assays ›
Gene Symbol:
CEP70
Gene Name:
centrosomal protein 70
Gene Aliases:
BITE
Chromosome Location:
Chr.3: 138494339 - 138594383 on Build GRCh38
UniGene ID:
Hs.531962
Species:
Human
siRNA ID
HSS129607
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP70
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001288964.1
NP_001275893.1
17
1975
NM_001288965.1
NP_001275894.1
16
1919
NM_001288967.1
NP_001275896.1
13
1523
NM_001320598.1
NP_001307527.1
17
2045
NM_001320599.1
NP_001307528.1
17
1993
NM_024491.3
NP_077817.2
17
1993
XM_005247805.3
XP_005247862.1
15
1713
XM_017007270.1
XP_016862759.1
15
1713
XM_017007271.1
XP_016862760.1
17
1895
XM_017007272.1
XP_016862761.1
17
2233
XM_017007273.1
XP_016862762.1
16
1838
XM_017007274.1
XP_016862763.1
16
2258
XM_017007275.1
XP_016862764.1
17
2329
XM_017007276.1
XP_016862765.1
13
1526
XM_017007277.1
XP_016862766.1
13
1458
XM_017007278.1
XP_016862767.1
18
2064
XM_017007279.1
XP_016862768.1
16
1827
XM_017007280.1
XP_016862769.1
15
1668
XM_017007281.1
XP_016862770.1
10
1841
XM_017007282.1
XP_016862771.1
16
1794
XM_017007283.1
XP_016862772.1
16
1725
XM_017007284.1
XP_016862773.1
10
2029
GenBank mRNA
AF202146.1
-
17
1847
AK023098.1
-
15
1708
AK294587.1
-
13
1398
AK299003.1
-
16
1784
AK301913.1
-
17
1852
AK314314.1
-
17
1965
BC030598.2
-
17
1895
4829481
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0