Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002019.2 | NP_001002019.1 | 4, 5 | 612 | ||
NM_001002020.2 | NP_001002020.1 | 4, 5 | 641 | |||
NM_025215.5 | NP_079491.2 | 4, 5 | 989 | |||
XM_011538768.2 | XP_011537070.1 | 3, 4 | 254 | |||
GenBank mRNA | AF116238.1 | - | 3, 4 | 349 | ||
AF318369.1 | - | 4, 5 | 981 | |||
AK074659.1 | - | 4, 5 | 989 | |||
AK292242.1 | - | 4, 5 | 585 | |||
BC002901.2 | - | 4, 5 | 555 | 3943243 | ||
BC009505.2 | - | 4, 5 | 760 | 2822709 | ||
BC019320.1 | - | 4, 5 | 760 | 2822709 | ||
BC035964.1 | - | 3, 4 | 1129 | |||
BM923133.1 | - | 4, 5 | 618 | |||
BX340929.2 | - | 4, 5 | 616 |