Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002019.2 | NP_001002019.1 | 5 | 647 | ||
NM_001002020.2 | NP_001002020.1 | 5 | 676 | |||
NM_025215.5 | NP_079491.2 | 5 | 1024 | |||
XM_011538768.2 | XP_011537070.1 | 4 | 289 | |||
GenBank mRNA | AF116238.1 | - | 4 | 384 | ||
AF318369.1 | - | 5 | 1016 | |||
AK074659.1 | - | 5 | 1024 | |||
AK292242.1 | - | 5 | 620 | |||
BC002901.2 | - | 5 | 590 | 3943243 | ||
BC009505.2 | - | 5 | 795 | 2822709 | ||
BC019320.1 | - | 5 | 795 | 2822709 | ||
BC035964.1 | - | 4 | 1164 | |||
BX340929.2 | - | 5 | 651 |