Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025216.2 | NP_079492.2 | 2 | 822 | ||
XM_011511929.2 | XP_011510231.1 | 3 | 430 | |||
XM_011511930.1 | XP_011510232.1 | 2 | 475 | |||
GenBank mRNA | AB059569.1 | - | 2 | 364 | ||
AK024363.1 | - | 2 | 822 | |||
AK315081.1 | - | 2 | 475 | |||
AY009400.1 | - | 2 | 555 | |||
BC034352.1 | - | 2 | 481 | |||
BC040026.1 | - | 2 | 792 | |||
BC052234.1 | - | 2 | 833 | 5442628 |