Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025227.2 | NP_079503.1 | 9, 10 | 1081 | ||
GenBank mRNA | AF465765.1 | - | 8, 9 | 864 | ||
AK027068.1 | - | 9, 10 | 1059 | |||
AK172819.1 | - | 9, 10 | 1060 | |||
AK304064.1 | - | 9, 10 | 911 | |||
AY358723.1 | - | 9, 10 | 989 | |||
BC034415.1 | - | 9, 10 | 935 | 5173032 |