Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025227.2 | NP_079503.1 | 13 | 1398 | ||
GenBank mRNA | AF465765.1 | - | 12 | 1181 | ||
AK027068.1 | - | 13 | 1376 | |||
AK172819.1 | - | 13 | 1377 | |||
AK304064.1 | - | 13 | 1228 | |||
AY358723.1 | - | 13 | 1306 | |||
BC034415.1 | - | 13 | 1252 | 5173032 |